Gold trichloride ≥99.99% (metals basis) Au ≥ 64.4%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Gold(III) chloride, Gold (III) chloride
43360-03
43360-06
CAAA43360-03EA
328.64
CAD
CAAA43360-03
CAAA43360-06
Gold trichloride ≥99.99% (metals basis) Au ≥ 64.4%, Premion®
Gold trichloride
Purity: 99.99%
CAS No.: 13453-07-1
Molecular Formula: AuCl3
Formula Weight: 303.33
Physical Form: Crystalline
Density: 3.90
Appearance: Red
Odor: Odorless
Melting Point: 180(subl)
Merck Reference: 14,4521
UN No.: 3260
MDL No.: MFCD00014172
Formula:
AuCl₃ MW: 303.32 g/mol Melting Pt: 180 °C (subl.) Density: 3.90 Storage Temperature: Ambient |
MDL Number:
MFCD00014172 CAS Number: 13453-07-1 EINECS: 236-623-1 UN: 3260 ADR: 8,III Merck Index: 14,4521 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...