Acrylic acid, low water content ≥99.5% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Prop-2-enoic acid

Stabilization: ∼200 ppm Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)

43359-AP 43359-K4
CAAA43359-APEA 87.04 CAD
CAAA43359-AP CAAA43359-K4
Acrylic acid, low water content ≥99.5% stabilised
Acrylic acid

MDL: MFCD00004367 Beilstein: 635743 Notes: Water less than 0.1%. Miscible with water, alcohols, ether


Purity: 99.5% CAS No.: 79-10-7 Molecular Formula: H2C=CHCO2H Formula Weight: 72.06 Flash Point: 54°(130°F) Physical Form: Liquid Density: 1.051 Refractive Index: 1.4202 Odor: Pungent Melting Point: ca 13° Boiling Point: 138-139° Solubility: Miscible with water, alcohols, ether Beilstein Reference: 635743 Merck Reference: 14,130 UN No.: 2218 MDL No.: MFCD00004367

Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 138…139 °C
Melting Pt: ∼ 13 °C
Density: 1.051
Flash Pt: 54 °C (130 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004367
CAS Number: 79-10-7
EINECS: 201-177-9
UN: 2218
ADR: 8 II
Merck Index: 14,130

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR