Magnesium 99,9 Metals basis, rods, Length: 25 mm (1 in), Ø 7.9 mm (0.31 in)

Supplier: Thermo Scientific Chemicals

43355-KF 43355-KJ 43355-KM
CAAA43355-KFEA 60.04 CAD
CAAA43355-KF CAAA43355-KJ CAAA43355-KM
Magnesium 99,9 Metals basis, rods, Length: 25 mm (1 in), Ø 7.9 mm (0.31 in)
Magnesium

Purity: 99.9% CAS No.: 7439-95-4 Physical Form: ≈2.1g/piece Odor: Odorless MDL No.: MFCD00085308

Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
Density: 1.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR