(±)-3-Hydroxybutyric acid, tech.

Supplier: Thermo Scientific Chemicals
Warning

43354-06 43354-14
CAAAA43354-06EA 181.7 CAD
CAAAA43354-06 CAAAA43354-14
(±)-3-Hydroxybutyric acid, tech.
(±)-3-Hydroxybutyric acid

MDL: MFCD00004546 Soluble in water, alcohol, ether


CAS No.: 300-85-6 Molecular Formula: C4H8O3 Formula Weight: 104.10 Flash Point: 112°(234°F) Physical Form: Liquid Density: 1.13 Refractive Index: 1.4445 Sensitivity: Hygroscopic Appearance: Colorless Boiling Point: 130°/12mm Solubility: Soluble in water, alcohol, ether Merck Reference: 14,4816 MDL No.: MFCD00004546

Formula: C₄H₈O₃
Boiling Pt: 130 °C (12 mmHg)
Melting Pt: 33…34 °C
Density: 1.13
Flash Pt: >112 °C (234 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004546
CAS Number: 300-85-6
EINECS: 206-099-9
Merck Index: 14,4816

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR