Nickel ≥99.8% (metals basis), spherical powder APS 0.08-0.15 µm

Supplier: Thermo Scientific Chemicals
Danger

43338-09 43338-18 43338-30
CAAA43338-09EA 164.32 CAD
CAAA43338-09 CAAA43338-18 CAAA43338-30
Nickel ≥99.8% (metals basis), spherical powder APS 0.08-0.15 µm
Nickel

Purity: 99.8% CAS No.: 7440-02-0 Physical Form: S.A. 4.5-8.5m2/g, Tap density 1.0-2.5g/cm3 Odor: Odorless UN No.: 3089 MDL No.: MFCD00011137

Formula: Ni
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR