Hydrochloric acid 5% (v/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

43335-AP 43335-K2 43335-K7
CAAAA43335-APEA 39.03 CAD
CAAAA43335-AP CAAAA43335-K2 CAAAA43335-K7
Hydrochloric acid 5% (v/v) in aqueous solution
Hydrochloric acid

Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colorless to pale yellow UN No.: 1789 MDL No.: MFCD00011324

Formula: HCl
MW: 36.46
Boiling Pt: ∼100 °C (1013 hPa)
Density: 1.02…1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR