Calcium chromate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

43333-14 43333-22
CAAA43333-14EA 152 CAD
CAAA43333-14 CAAA43333-22
Calcium chromate ≥99.9% (metals basis)
Calcium chromate

Purity: 99.9% CAS No.: 13765-19-0 Molecular Formula: CaCrO4 Formula Weight: 156.07 Physical Form: -20 Mesh Powder Appearance: Yellow UN No.: 3087 MDL No.: MFCD00049403

Formula: CaCrO₄
Melting Pt: 2170 °C
Density: 3.12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049403
CAS Number: 13765-19-0
EINECS: 237-366-8
UN: 3087
ADR: 5.1,III
Merck Index: 12,01700

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR