Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, REacton®, thickness 0.075 mm (0.003 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
43307-FF
43307-FI
CAAA43307-FFEA
165.9
CAD
CAAA43307-FF
CAAA43307-FI
Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, REacton®, thickness 0.075 mm (0.003 in)
Gadolinium
Purity: 99.9%
CAS No.: 7440-54-2
Physical Form: ≈0.36g/25x25mm, Packaged under argon
Appearance: Gray
UN No.: 3178
MDL No.: MFCD00011022
Formula:
Gd Storage Temperature: Ambient |
MDL Number:
MFCD00011022 CAS Number: 7440-54-2 EINECS: 231-162-2 UN: 3178 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...