(±)-1-Ethoxy-2-propanol 90+% remainder 2-ethoxy-1-propanol
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Ethoxy-2-propanol, (±)-Monopropylene glycol ethyl ether
43306-AP
43306-AY
CAAA43306-APEA
169.06
CAD
CAAA43306-AP
CAAA43306-AY
(±)-1-Ethoxy-2-propanol 90+% remainder 2-ethoxy-1-propanol
(±)-1-Ethoxy-2-propanol
Purity: 90+%
CAS No.: 1569-02-4
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 42°(108°F)
Physical Form: Liquid
Density: 0.896
Refractive Index: 1.4065
Appearance: Colorless
Melting Point: -100°
Boiling Point: 132°
UN No.: 1987
MDL No.: MFCD00067050
Formula:
C₅H₁₂O₂ Boiling Pt: 132 °C Melting Pt: –100 °C Density: 0.896 Flash Pt: 42 °C (108 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00067050 CAS Number: 1569-02-4 EINECS: 216-374-5 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...