n-Hexadecane ≥95%

Supplier: Thermo Scientific Chemicals
Danger

43283-LV 43283-LW
CAAA43283-LVEA 287.56 CAD
CAAA43283-LV CAAA43283-LW
n-Hexadecane ≥95%
n-Hexadecane

Standard for rating diesel-fuel ignition qualities


Purity: 95% CAS No.: 544-76-3 Molecular Formula: CH3(CH2)14CH3 Formula Weight: 226.45 Flash Point: 135°(275°F) Physical Form: Liquid Density: 0.773 Refractive Index: 1.4345 Appearance: Colorless Melting Point: 18° Boiling Point: 287° Solubility: Soluble in ethanol Beilstein Reference: 1736592 MDL No.: MFCD00008998


Standard for rating diesel-fuel ignition qualities

Formula: H₃C(CH₂)₁₄CH₃
Boiling Pt: 287 °C
Melting Pt: 18 °C
Density: 0.773
Flash Pt: 135 °C (275 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR