Silver(I) oxide electrical grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
43268-30
43268-A1
CAAA43268-30EA
1698.5
CAD
CAAA43268-30
CAAA43268-A1
Silver(I) oxide electrical grade
Silver(I) oxide
For battery plates, pollution control filters
CAS No.: 20667-12-3
Molecular Formula: Ag2O
Formula Weight: 231.74
Physical Form: -325 Mesh Powder
Density: 7.2
Sensitivity: Light Sensitive
Appearance: Dark brown
Odor: Odorless
Melting Point: 230° dec.
Solubility: Soluble in dilute HNO3 and ammonia. Practically insoluble in water. Insoluble in alcohol
Merck Reference: 14,8521
UN No.: 1479
MDL No.: MFCD00003404
For battery plates, pollution control filters
Formula:
Ag₂O Melting Pt: 230 °C (dec.) Density: 7.2 Storage Temperature: Ambient |
MDL Number:
MFCD00003404 CAS Number: 20667-12-3 EINECS: 243-957-1 UN: 1479 ADR: 5.1,II Merck Index: 14,8521 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...