2-Amino-4-methylbenzonitrile ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43260-03
43260-14
CAAAA43260-03EA
68.57
CAD
CAAAA43260-03
CAAAA43260-06
CAAAA43260-14
2-Amino-4-methylbenzonitrile ≥98%
2-Amino-4-methylbenzonitrile
Purity: 98%
CAS No.: 26830-96-6
Molecular Formula: C8H8N2
Formula Weight: 132.17
Physical Form: Powder
Melting Point: 91-93°
UN No.: 3439
MDL No.: MFCD00173706
Formula:
C₈H₈N₂ Melting Pt: 91…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173706 CAS Number: 26830-96-6 EINECS: 248-020-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...