Cobalt ≥99.995% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
43227-QI
43227-QJ
43227-QK
CAAA43227-QIEA
114.55
CAD
CAAA43227-QI
CAAA43227-QJ
CAAA43227-QK
Cobalt ≥99.995% (metals basis), slug, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Cobalt
Purity: 99.995%
CAS No.: 7440-48-4
Physical Form: ≈3.6g/slug
Odor: Odorless
MDL No.: MFCD00010935
Formula:
Co Boiling Pt: 2870 °C (1013 hPa) Melting Pt: 1495 °C Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...