Quinoline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43225-18
43225-30
43225-A1
CAAA43225-18EA
51.19
CAD
CAAA43225-18
CAAA43225-30
CAAA43225-A1
Quinoline 98%
Quinoline
Purity: 98%
CAS No.: 91-22-5
Molecular Formula: C9H7N
Formula Weight: 129.16
Flash Point: 101°(214°F)
Physical Form: Liquid
Density: 1.090
Refractive Index: 1.6260
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Colorless to pale yellow
pH: 7.3
Melting Point: -15°
Boiling Point: 237°
Beilstein Reference: 107477
Merck Reference: 14,8068
UN No.: 2656
MDL No.: MFCD00006736
Formula:
C₉H₇N Boiling Pt: 237 °C Melting Pt: –15 °C Density: 1.090 Flash Pt: 101 °C (214 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006736 CAS Number: 91-22-5 EINECS: 202-051-6 UN: 2656 ADR: 6.1,III Merck Index: 14,8068 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...