Nickel ≥99.8% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

43214-14 43214-22 43214-36 43214-A3
CAAA43214-14EA 37.6 CAD
CAAA43214-14 CAAA43214-22 CAAA43214-36 CAAA43214-A3
Nickel ≥99.8% (metals basis), powder -325 mesh
Nickel

Conductive surface coatings, inks, adhesives, and sealants


Purity: 99.8% CAS No.: 7440-02-0 Physical Form: Conductive dendritic powder Odor: Odorless UN No.: 3089 MDL No.: MFCD00011137


Conductive surface coatings, inks, adhesives, and sealants

Formula: Ni
Boiling Pt: 2371 °C (1013 hPa)
Melting Pt: 1455 °C
Density: 8.9 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR