Magnesium ≥99.9% (metals basis), foil, hard, Thickness: 0.05 mm (0.002 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
43202-FF
43202-FI
CAAA43202-FFEA
281.24
CAD
CAAA43202-FF
CAAA43202-FI
Magnesium ≥99.9% (metals basis), foil, hard, Thickness: 0.05 mm (0.002 in)
Magnesium
Sacrificial electrode for other material, deoxidizer for Cu
Purity: 99.9%
CAS No.: 7439-95-4
Physical Form: ≈0.05g/25x25mm, Temper: as rolled
Odor: Odorless
UN No.: 1869
MDL No.: MFCD00085308
Sacrificial electrode for other material, deoxidizer for Cu
Formula:
Mg Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...