Strontium nitrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

43193-14 43193-22
CAAA43193-14EA 347.6 CAD
CAAA43193-14 CAAA43193-22
Strontium nitrate ≥99.999% (metals basis), Puratronic®
Strontium nitrate

Purity: 99.999% CAS No.: 10042-76-9 Molecular Formula: N2O6Sr Formula Weight: 211.63 Physical Form: Crystalline, Packaged under argon Density: 2.990 Appearance: White Odor: Odorless Melting Point: 570° Merck Reference: 14,8845 UN No.: 1507 MDL No.: MFCD00011248

Formula: Sr(NO₃)₂
MW: 211.63 g/mol
Boiling Pt: 645
Melting Pt: 570
Density: 2.990
Storage Temperature: Ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1 III
Merck Index: 14,8845

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR