Ammonium hexafluorotitanate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ammonium fluotitanate, Ammonium hexafluorotitanate, Ammonium hexafluorotitanate(IV), Ammonium hexafluorotitanate (IV)

43192-14 43192-22 43192-36
CAAA43192-14EA 192.76 CAD
CAAA43192-14 CAAA43192-22 CAAA43192-36
Ammonium hexafluorotitanate ≥99.99% (metals basis)
Ammonium hexafluorotitanate

Purity: 99.99% CAS No.: 16962-40-6 Molecular Formula: (NH4)2TiF6 Formula Weight: 197.95 Physical Form: Crystalline Appearance: White MDL No.: MFCD00010888

Formula: H₈F₆N₂Ti
MW: 197.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010888
CAS Number: 16962-40-6
EINECS: 241-036-9
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR