Ammonium hexafluorosilicate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

43184-09 43184-18 43184-30
CAAA43184-09EA 101.12 CAD
CAAA43184-09 CAAA43184-18 CAAA43184-30
Ammonium hexafluorosilicate ≥99.999% (metals basis), Puratronic®
Ammonium hexafluorosilicate

Purity: 99.999% CAS No.: 16919-19-0 Molecular Formula: (NH4)2SiF6 Formula Weight: 178.14 Physical Form: Crystalline Melting Point: dec. UN No.: 2854 MDL No.: MFCD00010887

Formula: H₈F₆N₂Si
Melting Pt: Dec.
Density: 2.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010887
CAS Number: 16919-19-0
EINECS: 240-968-3
UN: 2854
ADR: 6.1,III
Merck Index: 14,527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR