Cerium(III) nitrate hydrate ≥99.998% (metals basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
43181-14
43181-22
CAAA43181-14EA
248.06
CAD
CAAA43181-14
CAAA43181-22
Cerium(III) nitrate hydrate ≥99.998% (metals basis), REacton®
Cerium trinitrate
Purity: 99.998%
Molecular Formula: Ce(NO3)3.xH2O (x~6-7)
Formula Weight: 326.13(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Odor: Acidic
Merck Reference: 14,2000
UN No.: 1477
MDL No.: MFCD00010926
Formula:
Ce(NO₃)₃·xH₂O MW: 326.13 Density: 2.377 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010926 CAS Number: 10108-73-3 EINECS: 233-297-2 UN: 1477 ADR: 5.1 III Merck Index: 14,2000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...