Ammonium hexafluorogermanate(IV) ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium hexafluorogermanate

43179-06 43179-14
CAAA43179-06EA 87.22 CAD
CAAA43179-06 CAAA43179-14
Ammonium hexafluorogermanate(IV) ≥99.99% (metals basis)
Ammonium hexafluorogermanate (IV)

Purity: 99.99% min CAS No.: 16962-47-3 Molecular Formula: H8F6GeN2 Formula Weight: 222.71 Physical Form: Crystalline Density: 2.564 Appearance: White UN No.: 3288 MDL No.: MFCD00010880

Formula: H₈F₆GeN₂
MW: 222.68 g/mol
Melting Pt: 380 °C
Density: 2.564
Storage Temperature: Ambient
MDL Number: MFCD00010880
CAS Number: 16962-47-3
EINECS: 241-037-4
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR