Triethylgermane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
43176-03
43176-06
CAAA43176-03EA
154.68
CAD
CAAA43176-03
CAAA43176-06
Triethylgermane 97%
Co-reactant for the preparation of germanium acrylates
Purity: 97%
CAS No.: 1188-14-3
Molecular Formula: (C2H5)3GeH
Formula Weight: 160.78
Flash Point: 5°(41°F)
Physical Form: Liquid
Density: 1.004
Refractive Index: 1.4382
Boiling Point: 122°
UN No.: 3399
MDL No.: MFCD00077989
Co-reactant for the preparation of germanium acrylates
Formula:
C₆H₁₆Ge Boiling Pt: 122 °C Density: 1.004 Flash Pt: 5 °C (41 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00077989 CAS Number: 1188-14-3 UN: 3399 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...