2-Ethylhexyl methacrylate ≥98% stabilised

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methacrylic acid 2-ethylhexyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

42303-AK 42303-K2 42303-K7 42303-M4
CAAA42303-AKEA 47.56 CAD
CAAA42303-AK CAAA42303-K2 CAAA42303-K7 CAAA42303-M4
2-Ethylhexyl methacrylate ≥98% stabilised
2-Ethylhexyl methacrylate

Monomer used in the preparation of polymethacrylate polymers


Purity: 98% CAS No.: 688-84-6 Molecular Formula: C12H22O2 Formula Weight: 198.31 Flash Point: 92°(198°F) Physical Form: Liquid Density: 0.885 Refractive Index: 1.4380 Appearance: Colorless Odor: Ether-like Melting Point: -50° Boiling Point: 218° Beilstein Reference: 1769420 MDL No.: MFCD00009494


Monomer used in the preparation of polymethacrylate polymers

Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
Boiling Pt: 218 °C
Melting Pt: –50 °C
Density: 0.885
Flash Pt: 92 °C (198 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009494
CAS Number: 688-84-6
EINECS: 211-708-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR