Tin 99,995 Metals basis, slug, Puratronic®, Ø 11 mm (0.43 in), Thickness: 2 mm (0.1 in)

Supplier: Thermo Scientific Chemicals

41169-KD 41169-KK
CAAA-41169-KDEA 1215.02 CAD
CAAA-41169-KD CAAA-41169-KK
Tin 99,995 Metals basis, slug, Puratronic®, Ø 11 mm (0.43 in), Thickness: 2 mm (0.1 in)
Tin

Evaporation source


Purity: 99.995% CAS No.: 7440-31-5 Physical Form: ≈1.3g Odor: Odorless MDL No.: MFCD00133862


Evaporation source

Formula: Sn
MW: 118.71 g/mol
Boiling Pt: 2270
Melting Pt: 232
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR