Rose's metal (Bi:Pb:Sn 50:25:25 wt%), ingot
Supplier: Thermo Scientific Chemicals
Danger
40516-22 40516-36
CAAA40516-22EA 263.86 CAD
CAAA40516-22 CAAA40516-36
Rose's metal (Bi:Pb:Sn 50:25:25 wt%), ingot
Rose's metal
Molecular Formula: Bi:Pb:Sn; 50:25:25 wt% Physical Form: Low melting alloy Melting Point: 95-96° MDL No.: MFCD00804027