Erbium triacetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

Synonyms: Erbium(III) acetate tetrahydrate

40481-06 40481-14 40481-22
CAAA40481-06EA 79.47 CAD
CAAA40481-06 CAAA40481-14 CAAA40481-22
Erbium triacetate tetrahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium triacetate tetrahydrate

Purity: 99.9% CAS No.: 15280-57-6 Molecular Formula: C6H9ErO6.4H2O Formula Weight: 416.45 (344.39anhy) Physical Form: Crystalline Aggregates Density: 2.114 MDL No.: MFCD00013034

Formula: C₆H₉ErO₆·4H₂O
MW: 416.45 g/mol
Density: 2,114
Storage Temperature: Ambient
MDL Number: MFCD00013034
CAS Number: 15280-57-6
EINECS: 247-067-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR