Thallium(I) bromide, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
35720-09
35720-18
CAAA35720-09EA
183.28
CAD
CAAA35720-09
CAAA35720-18
Thallium(I) bromide, ultra dry ≥99.998% (metals basis)
Thallium (I) bromide
Purity: 99.998%
CAS No.: 7789-40-4
Molecular Formula: TlBr
Formula Weight: 284.28
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 7.5
Melting Point: ca 460°
Merck Reference: 14,9258
UN No.: 1707
MDL No.: MFCD00011273
Formula:
BrTl Boiling Pt: 815 °C (1013 hPa) Melting Pt: ∼460 °C Density: 7.5 Storage Temperature: Ambient |
MDL Number:
MFCD00011273 CAS Number: 7789-40-4 EINECS: 232-163-0 UN: 1707 ADR: 6.1,II Merck Index: 14,9258 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...