Ytterbium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
Synonyms:
Ytterbium(III) acetate hydrate
14576-09
14576-18
CAAA14576-09EA
98.91
CAD
CAAA14576-09
CAAA14576-18
Ytterbium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Ytterbium triacetate hydrate
Purity: 99.9%
CAS No.: 15280-58-7
Molecular Formula: Yb(OOCCH3)3.xH2O
Formula Weight: 350.16(anhy)
Physical Form: Crystalline Aggregates
Odor: Odorless
MDL No.: MFCD00150127
Formula:
C₆H₉O₆Yb·H₂O MW: 368.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150127 CAS Number: 15280-58-7 EINECS: 244-137-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...