Silver trifluoroacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

13933-06 13933-14
CAAA13933-06EA 98.43 CAD
CAAA13933-06 CAAA13933-14
Silver trifluoroacetate 98%
Silver trifluoroacetate

Purity: 98% CAS No.: 2966-50-9 Molecular Formula: C2AgF3O2 Formula Weight: 220.88 Physical Form: Powder Sensitivity: Light Sensitive and Hygroscopic Appearance: White to grey Odor: Odorless Melting Point: 257-260° dec. UN No.: 3077 MDL No.: MFCD00013199

Formula: C₂AgF₃O₂
Melting Pt: 257…260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013199
CAS Number: 2966-50-9
EINECS: 221-004-0
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR