Potassium osmate(VI) dihydrate ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
12647-01
12647-03
CAAA12647-01EA
120.87
CAD
CAAA12647-01
CAAA12647-03
Potassium osmate(VI) dihydrate ≥98%
Potassium osmate(VI) dihydrate
Purity: 98+%
CAS No.: 10022-66-9
Molecular Formula: K2OsO4.2H2O
Formula Weight: 368.43 (332.40anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Purple
Solubility: Soluble in water. Slowly decomposes in aqueous solution forming the tetroxide. Insoluble in alcohol, ether
UN No.: 3288
MDL No.: MFCD00149919
Formula:
K₂O₄Os·2H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149919 CAS Number: 10022-66-9 EINECS: 243-247-1 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...