Zinc(II) acetylacetonate monohydrate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Zinc 2,4-pentanedionate monohydrate, Zinc 2,4-pentanedionate hydrate, Bis(2,4-pentanedionato-O,O')zinc (II) monohydrate, Zinc (II) bis(2,4-pentanedionato-O,O') monohydrate, Zinc (II) 2,4-pentanedionate monohydrate

12527-18 12527-30
CAAA12527-18EA 61.94 CAD
CAAA12527-18 CAAA12527-30
Zinc(II) acetylacetonate monohydrate
Zinc (II) acetylacetonate monohydrate

MDL: MFCD00000035 Soluble in alcohol. Slightly soluble in water


CAS No.: 14363-15-6 Molecular Formula: C10H14O4Zn.H2O Formula Weight: 281.60 (263.59anhy) Physical Form: Crystalline Density: 1.6 Melting Point: 136-138° dec. MDL No.: MFCD00000035

Formula: C₁₀H₁₄O₄Zn·1H₂O
MW: 281.62 g/mol
Melting Pt: 136…138 °C (dec.)
Density: 1.6
Storage Temperature: Ambient
MDL Number: MFCD00000035
CAS Number: 14363-15-6
EINECS: 237-860-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR