Palladium (II) sulfate dihydrate ≥99.95% (metals basis) Pd ≥ 44.1%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
12165-01
12165-06
CAAA12165-01EA
67.94
CAD
CAAA12165-01
CAAA12165-06
Palladium (II) sulfate dihydrate ≥99.95% (metals basis) Pd ≥ 44.1%, Premion®
Palladium (II) sulphate dihydrate
Purity: 99.95%
CAS No.: 13444-98-9
Molecular Formula: O4PdS.2H2O
Formula Weight: 238.51 (202.48anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Red to brown
Melting Point: dec.
UN No.: 3260
MDL No.: MFCD00011173
Formula:
O₄PdS·2H₂O Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00011173 CAS Number: 13444-98-9 EINECS: 236-957-8 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...