Ammonium hexachlororuthenate(IV) Ru ≥ 28,4%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diammonium hexachlororuthenate

11874-03 11874-06
CAAA11874-03EA 102.54 CAD
CAAA11874-03 CAAA11874-06
Ammonium hexachlororuthenate(IV) Ru ≥ 28,4%
Ammonium hexachlororuthenate(IV)

CAS No.: 18746-63-9 Molecular Formula: (NH4)2RuCl6 Formula Weight: 349.87 Physical Form: Crystalline powder Sensitivity: Hygroscopic Appearance: Red-brown Odor: Odorless MDL No.: MFCD00015962

Formula: H₈Cl₆N₂Ru
MW: 349.86
Storage Temperature: Ambient
MDL Number: MFCD00015962
CAS Number: 18746-63-9
EINECS: 242-552-7
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR