Zinc iodide 98%

Supplier: Thermo Scientific Chemicals
Danger

11661-18 11661-30
CAAA11661-18EA 129.72 CAD
CAAA11661-18 CAAA11661-30
Zinc iodide 98%
Zinc iodide

MDL: MFCD00011299 Freely soluble in water, glycerol, alcohol, ether

Formula: I₂Zn
Boiling Pt: 625 °C (dec.)
Melting Pt: 446 °C
Density: 4.74
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS Number: 10139-47-6
EINECS: 233-396-0
UN: 3260
ADR: 8,II
Merck Index: 14,10140

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR