Removable Small Hub Needles for Syringes 2.5 μl to 10 ml, Hamilton Company
Supplier: Hamilton
7762-01 7762-02 7762-03 7762-04 7762-06 7758-01 7758-02 7758-03 7758-04 7768-02 7784-08 7784-07 7784-06 7784-05 7770-02 7770-01 7762-05 7768-01 7803-01 7803-05 8646-03
82010-224PK 503.01 CAD
82010-224 82010-226 82010-228 82010-230 82010-234 82010-236 82010-238 82010-240 82010-242 82010-246 82010-248 82010-250 82010-252 82010-254 82010-258 82007-208 82010-232 82010-244 89210-098 89221-012 10746-556
Removable Small Hub Needles for Syringes 2.5 μl to 10 ml, Hamilton Company
Syringe Needles
Hamilton Company offers the most comprehensive selection of standard and custom needles in the industry. Each is manufactured to achieve the highest level of accuracy and precision possible.
Removable Needle (RN)- The needles are removable and are a Hamilton-specific design.
Needle Point Styles (Custom gauges, lengths and point styles available):
Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34-10.
Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 34-10.
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable