PTFE Tubing Adapted to Female Luer Lock Fitting, Hamilton Company
Supplier: Hamilton
90618 90619 90620 90622 90626 90630 90611 90616 90617 90668 90662 90607 90608 90609 90610 90613 90614 90615 90624 90628 90641
60377-506EA 66.65 CAD
60377-506 60377-508 60377-510 60377-512 60377-516 60377-520 60377-492 60377-502 60377-504 89217-244 89217-246 89221-504 89221-506 89221-508 89221-510 89221-514 89221-516 89221-518 89221-520 89221-522 89221-524
PTFE Tubing Adapted to Female Luer Lock Fitting, Hamilton Company
Tubing Transfer Tubing
Hamilton Company offers the most comprehensive selection of standard and custom syringes, valves and accessories in the industry.
This tubing comes with a female luer lock connection on one end and has a raw cut on the opposite end. The tubing is 12 inches in length but can be trimmed shorter to a desired length. The tubing is available in sizes from 30 gauge to 7 gauge.
Each is manufactured to achieve the highest level of accuracy and precision possible.
Order Now
SPECIFICATIONS
Environmentally Preferable