Microliter™ 800 Series Syringe Replacement Barrel/Plunger Assembly, Hamilton Company
Supplier: Hamilton
32117 32129 32151 32165
60373-225EA 240.83 CAD
60373-225 60373-233 72310-432 89221-398
Microliter™ 800 Series Syringe Replacement Barrel/Plunger Assembly, Hamilton Company
Syringe Barrels, Plungers and Pistons
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry.
These replacement syringe barrel/plunger assemblies are for Hamilton 800 series Microliter™ syringes. The barrels range in volume from 25 to 250 µl with removable needle (RN) and cemented needle (N) terminations.
Each is manufactured to achieve the highest level of accuracy and precision possible.
Note: The picture shows the entire 800 series Microliter™ syringe. The product mentioned here is only the barrel/plunger assembly.
Hamilton Model 801 N replacement barrel/plungerMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable