Nalgene® Packaging Bottles, Narrow Mouth, HDPE, Amber, without Closures, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
362085-0001 362085-0004
16060-050CS 941.68 CAD
16060-050 16060-054
Nalgene® Packaging Bottles, Narrow Mouth, HDPE, Amber, without Closures, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene® narrow-mouth amber bottles are high density polyethylene and are leak proof.
These bottles meet the USP requirements of light-resistant containers. These opaque amber HDPE bottles are without closures.
Ordering information: For closures, see 16060-016 series.