Nalgene® IP2 Narrow Mouth Bottles, Natural HDPE, with PP Closure, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
312099-0001 312099-0002 312099-0004 312099-0008 312099-0016 312099-0032
16059-714CS 2186.06 CAD
16059-714 16059-716 16059-718 16059-720 16059-722 16059-724
Nalgene® IP2 Narrow Mouth Bottles, Natural HDPE, with PP Closure, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene® IP2 high density polyethylene narrow mouth bottles are recommended for storage and shipment of liquid reagents.
These bottles come with polypropylene closures and are recommended for customers involved in designing, assembling and certifying their own combination packaging. The bottles are safe, economical, and comply with the IATA and ICAO regulations.
Packaging: Bottles and closures are bulk packed in separate polybags within the same carton.