384-Well Non-Binding Microplate, Nonsterile, Greiner Bio-One
Supplier: Greiner Bio-One
781900 784900 781904 784904 781903 781906 781901
89089-584CS 398.2 CAD
89089-584 89089-586 89130-326 89130-328 89131-682 89131-684 89217-612
384-Well Non-Binding Microplate, Nonsterile, Greiner Bio-One
Microplates Deep Well Microplates
Greiner Bio-One's non-binding microplate surfaces prevent unwanted non-specific binding.
  • Nonsterile
  • Free of RNase, DNase, and DNA-olymerase inhibitors
  • Nonpyrogenic
  • Include solid and film well bottoms
  • Improved signal-to-noise ratio
This is especially advantageous for sensitive biochemical assays
All plates are RNase-, DNase-, and pyrogen-free, and also free of DNA-polymerase inhibitors.
Item numbers 89130-328 and 89089-586 come in HiBase plate design.
Low amounts of biomolecular binding can cause an undesirable increase in background, resulting in decreased signal-to-noise ratio. Characterized by low protein, DNA, RNA, and peptide binding properties, the non-binding microplate surfaces significantly increase assay sensitivity by reducing background and improving signal-to-noise ratio. Non-binding microplates are featured in 96 well and 384 well formats in black, white, and clear. Microplates include solid and µClear film well bottoms.
Ordering information: Lid not included.