Nalgene® Colored CryoBoxes, Thermo Scientific
Supplier: Thermo Fisher Scientific
867013-0240 867013-0241 867013-0242 867013-0243 867013-0244 867013-0245
73520-488CS 671.35 CAD
73520-488 73520-490 73520-492 73520-494 73520-496 73520-498
Nalgene® Colored CryoBoxes, Thermo Scientific
Boxes Cryoboxes
Nalgene® Colored CryoBoxes come in 5 grid colors for easy-to-identify inventory management, providing safe and economical storage of up to 81 cryogenic samples.
These colored boxes hold 1.0 to 2.0 ml cryogenic storage vials in a 9 x 9 array, and feature a usable temperature range of –196°C to +121°C. The colors match popular NALGENE cryogenic vial color coders.
The transparent lid allows easy viewing of contents and the lids have a numbered grid for quick sample identification.