FOCUS™ Proteome Kits, G-Biosciences
Supplier: G-Biosciences
786-360 786-246 786-257 786-258 786-259
CA78000-048EA 1321.91 CAD
CA78000-048 CA82022-670 CA82022-700 CA82022-702 CA82022-704
FOCUS™ Proteome Kits, G-Biosciences
Protein Extraction Protein Extraction Kits
An effective proteome analysis requires the preparation of a sample to bring the wide range of protein species into the dynamic range of detection
G-Biosciences' FOCUS™ Proteome Kits are for the preparation of total protein, including soluble, insoluble, membrane, cytoplasmic, nuclear, signal, phospho- and glyco-proteins.
G-Biosciences' FOCUS™ Proteome Kits are suitable for biological samples from tissues, cells, plants, yeast, bacteria and insects. The kits are simple to use, save time, improve the quality of protein analysis and enhance the chances of discovery of novel proteins.
G-Biosciences' FOCUS™ Proteome Kits are suitable for the analysis of proteins using electrophoresis and other biochemical techniques.
The absence of any standardized procedures for sample preparation has made proteome analysis extremely complicated, requiring a multitude of complicated skills, expensive equipment and resources.