CB™ Protein Assay, G-Biosciences
Supplier: G-Biosciences
786-012 786-012T
CA82022-366EA 529.74 CAD
CA82022-366 CA71003-086
CB™ Protein Assay, G-Biosciences
Assays Protein Assays
It is an improved Coomassie Dye based protein assay based on the Bradford Protein Assay
CB™ Protein Assay is supplied with a simple to follow protocol and a ready to use reagent that does not require prefiltering or dilution. Simply mix the protein solution with CB™ Protein Dye and read optical density.
The protein-dye complexes reach a stable end point in 5 minutes. The CB™ Protein Assay is compatible with reducing agents and a wide variety of common laboratory agents. Note: The Coomassie dye based assay is not suitable if the protein solution contains higher than recommended concentration of detergents or other agents. CB™ Protein Assay is sensitive over the linear range of 0.5-50 µg. Suitable for 500 assays.
This assay is suitable for the simple and rapid estimation of protein concentration. This assay is based on a single Coomassie dye based reagent. The binding of protein to the dye results in a change of color from brown to blue. The change in color density is proportional to protein concentration. Protein estimation can be performed using as little as 0.5 µg protein.
Order Now
SPECIFICATIONS
Environmentally Preferable