AlbuminOUT™, G-Biosciences
Supplier: G-Biosciences
786-251 786-252 786-251T
CA82022-680EA 1457.34 CAD
CA82022-680 CA82022-682 CA95057-554
AlbuminOUT™, G-Biosciences
Protein Extraction Protein Extraction Kits
G-Biosciences' AlbuminOUT™ has been specifically developed for substantial removal of albumin from samples that contain a large abundance of albumin, such as plasma and cerebrospinal fluid
G-Biosciences' AlbuminOUT™ albumin removal method is based on binding of albumin with Cibachron™ Blue dye. AlbuminOUT™ has been optimized for removal of human albumin from samples. AlbuminOUT™ uses a rapid spin column method, where each column contains 0.2 mL dye bound resins with capacity of >2 mg human albumin per column. AlbuminOUT™ will remove >98% albumin from 5-50 μL human plasma.
G-Biosciences' AlbuminOUT™ spin column format allows removal of albumin within 10 minutes. High capacity blue-dye binding resin allows instantaneous binding and removal of albumin from human, pig, sheep, dog, rabbit, rat, and bovine samples. AlbuminOUT™ may also be used for removal of albumin from other species. Suitable for processing 25 or 50 samples.
These samples tend to mask identification and discovery of other less abundant proteins in 2D gel electrophoresis and other studies.