Nalgene® Bulk-Packed, Non-Sterile, Cryogenic Vials, Thermo Scientific
Supplier: Thermo Fisher Scientific
5011-0012 5011-0020
66008-745CS 1061.26 CAD
66008-745 66008-748
Nalgene® Bulk-Packed, Non-Sterile, Cryogenic Vials, Thermo Scientific
Vials
Nalgene cryogenic vials are externally threaded vial for aseptic technique, using a design similiar to the linerless closures used on Nalgene bottles and carboys.
  • Self-standing and may be used without a holder
They are an economical, non-sterile version of Cat. Nos. 5000-0012 and -0020 without printing or graduations.
Ordering information: Closures and vials are packed separately.
Caution: For research use only; not for in-vitro diagnosis or parenterals. Do not autoclave.