Nalgene® Graduated Square Bottles, PC, Tray Packed, without Closures, Thermo Scientific
Supplier: Thermo Fisher Scientific
362015-0125 362015-0250 362015-0500 362015-1000
73521-076CS 695.64 CAD
73521-076 73521-078 73521-080 73521-082
Nalgene® Graduated Square Bottles, PC, Tray Packed, without Closures, Thermo Scientific
Bottles Plastic Bottles
Nalgene® graduated square bottles are polycarbonate, transparent, and autoclavable.
They have an easy-to-handle square design and molded-in graduations with a drip-proof neck. These bottles are ideal for storage of aqueous solutions and short-term storage of media.
Ordering information: These bottles are packaged in bulk. The number of bottles in the package depends upon the size of the bottle.