OPTI-SOLV® Micro Filters
Supplier: Optimize
10-04-03389 10-04-03621 10-04-03936 10-04-03707 10-04-03357
89042-066PK 1206.79 CAD
89042-066 89042-068 97056-520 89042-064 89209-704
OPTI-SOLV® Micro Filters
Filters Precolumn Filters and Frits
For volume critical filtering needs, optimize offers the OPTI-SOLV micro filter.
  • Hand-tight no tools required
  • Rated to 6000 psi
  • Designed to filter out particles to extend column life
  • Perfect Zero-Dead-Volume connections every time
  • Floating stem automatically adjusts to any depth port
Based on the design that made the mini filter successful, the micro filter cuts the internal volume to less than 200 nl while retaining the ease of use and functionality of the mini filter. The micro filter is designed to be used in today's low volume, high sensitivity chromatographic applications as a last line of defense against debris. As with all optimize column protection products, the OPTI-SOLV micro filter features a zero-dead volume connection utilizing patented floating stem (auto-adjusting) technology. Perfect zero-dead-volume connections are obtained every time with the patented automatic tube stop depth adjustment. The efficient design requires no holder and threads directly into any 10-32 port. The OPTI-SOLV mini, micro and nano filters are easy to use and perform exceptionally well for volume-critical applications.
Delivery information: Includes five disposable cartridges.