BRAND QuikSip™ Bottletop Fluid Aspiration System
Supplier: Brandtech
704526 6788
12777-154EA 53.95 CAD
12777-154 12777-184 89400-166
BRAND QuikSip™ Bottletop Fluid Aspiration System
Pumps Vacuum Aspiration Systems
The QuikSip™ is a convenient bottletop aspirator that removes supernatants, cell culture media, and other liquids from dishes, flasks, culture bottles, centrifugation tubes, multiwell plates, and more.
  • Economical spring powered aspirator
  • Fits 45 mm, 38 mm and 33 mm bottles (bottle not included)
  • Optional 8-channel manifold
The QuikSip™ works without any external vacuum source, allowing it to be easily transferred between workstations. Each plunger stroke powers up to 25mL of aspiration. The QuikSip™ includes a unique thumb-operated controller that isMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable