Nalgene® 50 Platinum-Cured Silicone Tubing for Peristaltic Pumps, Thermo Scientific
Supplier: Thermo Fisher Scientific
8060-3017 8060-3018 8060-3015 8060-3173 8060-3182 8060-3016 8060-3025 8060-3024
63015-318CS 479.48 CAD
63015-318 63015-320 63015-322 63015-328 63015-330 63015-314 63015-316 63015-324
Nalgene® 50 Platinum-Cured Silicone Tubing for Peristaltic Pumps, Thermo Scientific
Tubing Transfer Tubing
Flexible, durable, and translucent Platinum Silicone tubing is designed to provide extended pump life in peristaltic applications.
  • Tubing contains no phthalate plasticizers or peroxides that can leach out into transported fluid
  • Autoclavable
This high-purity tubing enables a variety of pump and transfer applications including pharmaceutical, laboratory, bioprocess manufacturing, and food and beverage.
Certifications: Tubing meets FDA, USDA, U.S. Pharmacopeia Class VI, 24th Edition; and 3A sanitary standards.
Ordering information: Supplied in 7.6m (25') coil lengths.
Caution: Note: Not intended for invasive use.