EXP® Fittings for 10-32 Ports and ¹/₁₆" Stainless Steel Tubing
Supplier: Optimize
15-20-03834 15-20-03842 15-20-03930 15-24-03832 15-20-03833 15-20-04120 15-20-03839 15-20-03841 15-20-03888 15-20-04128 15-24-03831 15-20-03840
97056-070PK 1678.2 CAD
97056-070 97056-074 97056-076 97056-078 97056-150 97056-152 97056-154 97056-156 97056-158 97056-160 97056-162 89208-082
EXP® Fittings for 10-32 Ports and ¹/₁₆" Stainless Steel Tubing
Fittings and Connectors
The EXP Fitting System is the premier adjustable nut and ferrule compression fitting for extreme high-pressure connections between ¹/₃₂ or ¹/₁₆" tubing and any 6-32, 6-40 or 10-32 port.
  • Rated to 20000+ psi (1400+ bar)
  • Tested to 30000 psi
  • Auto-adjusting ZDV Connection
  • Intended for many repeat uses
  • No more cut-off and replacement of swaged ferrules
  • Simple and versatile - can be used in UHPLC and HPLC applications
  • Use with EXP® Nuts and Holders
The patented titanium hybrid ferrule integrates the resilience of titanium with the proven sealing ability of PEEK in a superb one-piece design. The hybrid ferrule provides a perfect seal with every connection. The swage can be released without tools to adjust to the different port depths of various hardware. This adjustability provides value through repeated zero-dead-volume connections with any port configuration.